Learn More
Epredia™ Lab Vision™ Serotonin Ab-1, Mouse Monoclonal Antibody
Employ one marker in a comprehensive antibody portfolio that addresses all major disease states with the Epredia™ Serotonin Ab-1, Mouse Monoclonal Antibody.
Marca: Epredia™ MS-1431-S0
Detalles adicionales : Peso : 0.00170kg
Descripción
Serotonin-positive cells are present in gastric, duodenal, jejunal, ileal, colonic, and appendiceal mucosae. Positive cells are sparse and usually occur in the lower half of the gastric and intestinal mucosae. In the appendix, in addition to the mucosal serotonin-containing cells, there are single or small clusters of serotonin-positive cells in the sub-epithelial region of the lamina propria.Host Species: Mouse
Clone: 5HT-H209
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: 5-Hydroxytryptamine hydrochloride [3-(2-aminoethyl)-5-hydroxyindole hydrochloride; serotonin hydrochloride]
Positive Control: Appendix
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Especificaciones
Serotonin Ab-1 | |
Monoclonal | |
Unconjugated | |
5-Hydroxytryptamine hydrochloride (3-(2-aminoethyl)-5-hydroxyindole;serotonin) | |
100 μL | |
Neuroscience | |
Human | |
IgG2a κ |
Immunohistochemistry (Paraffin) | |
5HT-H209 | |
Mouse | |
Appendix | |
IVD | |
Primary | |
Concentrated |