missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ S100A4 Ab-8 Rabbit Polyclonal Antibody with BSA and azide

Epredia™ S100 A4 Ab-8, Rabbit Polyclonal Antibody provides accurate, reproducible results.

Marca:  Epredia™ RB-1804-R7

Detalles adicionales : Peso : 0.01200kg

 Ver más versiones de este producto

Código de producto. 12613157

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas


Descripción

Descripción

The S100A4 protein belongs to the S100 subfamily, now designated S100A1/S100A13. This antibody does not cross react with other S100 proteins. Anti-S100A4 labels fibroblasts, neuronal structures, T-lymphocytes, plasma cells and Kupffer cells in normal tissues.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse, Rat, Cow, Horse, Cat and Dog. Others not known.

Epitope: Not determined

Immunogen: Purified recombinant human S100 A4 monomer expressed in E. coli

Molecular Weight: 10-12kDa

Positive Control: Tonsil

Cellular Localization: Cytoplasmic, membrane and nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Especificaciones

Especificaciones

S100 A4 Ab-8
Polyclonal
Unconjugated
Purified recombinant human S100 A4 monomer expressed in E. coli
RUO
Primary
Prediluted
Immunohistochemistry (Paraffin)
Polyclonal
Rabbit
7 mL
Cancer and Tumor Biology
Bovine, Canine, Equine, Feline, Human, Mouse
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales