missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ p57Kip2 Ab-7 Rabbit Polyclonal Antibody with BSA and azide

Deliver accurate, reproducible results with the Epredia™ p57Kip2 Ab-7, Rabbit Polyclonal Antibody.

Marca:  Epredia™ RB-1637-P

Detalles adicionales : Peso : 0.00100kg

 Ver más versiones de este producto

Código de producto. 12653087

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas


Descripción

Descripción

p57Kip2 (or CDKN1C) is a potent tight-binding inhibitor of several G1 cyclin complexes, and is a negative regulator of cell proliferation. The gene encoding human p57Kip2 is located on chromosome 11p15.5, a region implicated in both sporadic cancers, Wilm's tumor, and Beckwith-Wiedemann syndrome (BWS), a cancer syndrome, making it a tumor suppressor candidate. BWS is characterized by numerous growth abnormalities and an increased risk of childhood tumors. Several types of childhood tumors including Wilms' tumor, adrenocortical carcinoma and rhabdomyosarcoma display a specific loss of maternal 11p15 alleles, suggesting that genomic imprinting plays an important part. This region also contains two other imprinted genes, insulin-like growth factor II (IGF-II) and H19, both of which seem to be implicated in adrenal neoplasms.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Cow and Mouse. Others not known.

Epitope: Near C-terminus

Immunogen: A synthetic peptide from human p57Kip2 protein

Molecular Weight: 57kDa

Positive Control: Colon CA or placenta

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not suitable)

Note:

USA: RUO; Int'l: RUO

Especificaciones

Especificaciones

p57superscriptKip2superscript Ab-7
Polyclonal
1mg/mL
Rabbit
1 mL
Cancer and Tumor Biology
Bovine, Human
IgG
Immunohistochemistry (Paraffin)
Polyclonal
Unconjugated
A synthetic peptide from human p57superscriptKip2superscript protein
RUO
Primary
Concentrated
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales