missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ p21WAF1 Ab-5 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Achieve accurate, reproducible results with the Epredia™ p21WAF1 Ab-5, Mouse Monoclonal Antibody.

Marca:  Epredia™ MS-387-PABX

Detalles adicionales : Peso : 0.00020kg

 Ver más versiones de este producto

Código de producto. 12651277

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas


Descripción

Descripción

p21WAF1/Cip1/Sdi1/Pic1 is a tumor suppressor protein. Expression of p21WAF1 is induced by wild type, but not mutant, p53 suppressor protein. The p21WAF1 protein binds to cyclin/CDK complexes and inhibits their kinase activity thereby stopping cell cycle progression. It also binds to PCNA (proliferating cell nuclear antigen) and blocks DNA replication but not the DNA repair process.

Host Species: Mouse

Clone: HZ52

Isotype: IgG1

Species Reactivity: Human, Cow, Mouse, and Rat. Others not known.

Epitope: Not determined

Immunogen: Full-length human recombinant p21 protein

Molecular Weight: 21kDa

Positive Control: Ls174T or Raji cells, Colon CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunoprecipitation
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Especificaciones

Especificaciones

p21superscriptWAF1superscript Ab-5
Monoclonal
1mg/mL
Mouse
200 μL
Cancer and Tumor Biology
Bovine, Human, Rat
IgG1
Immunohistochemistry (Paraffin), Immunoprecipitation
HZ52
Unconjugated
Full length human recombinant p21 protein
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers