Learn More
Epredia™ Lab Vision™ p21WAF1 Ab-5 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide
Achieve accurate, reproducible results with the Epredia™ p21WAF1 Ab-5, Mouse Monoclonal Antibody.
Marca: Epredia™ MS-387-P0
Detalles adicionales : Peso : 4.98950kg
Descripción
p21WAF1/Cip1/Sdi1/Pic1 is a tumor suppressor protein. Expression of p21WAF1 is induced by wild type, but not mutant, p53 suppressor protein. The p21WAF1 protein binds to cyclin/CDK complexes and inhibits their kinase activity thereby stopping cell cycle progression. It also binds to PCNA (proliferating cell nuclear antigen) and blocks DNA replication but not the DNA repair process.Host Species: Mouse
Clone: HZ52
Isotype: IgG1
Species Reactivity: Human, Cow, Mouse, and Rat. Others not known.
Epitope: Not determined
Immunogen: Full-length human recombinant p21 protein
Molecular Weight: 21kDa
Positive Control: Ls174T or Raji cells, Colon CA
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Immunoprecipitation
- Flow Cytometry
Note:
USA: RUO; Int'l: RUO
Especificaciones
p21superscriptWAF1superscript Ab-5 | |
Monoclonal | |
200μg/mL | |
Mouse | |
100 μL | |
Cancer and Tumor Biology | |
Bovine, Human, Rat | |
IgG1 |
Immunohistochemistry (Paraffin), Immunoprecipitation | |
HZ52 | |
Unconjugated | |
Full length human recombinant p21 protein | |
RUO | |
Primary | |
Concentrated |