missing translation for 'onlineSavingsMsg'
Learn More

p170/p-Glycoprotein/MDR Ab-2 Mouse Monoclonal Antibody, Epredia™

Ensure accurate, reproducible results with the Epredia™ p170/p-Glycoprotein/MDR Ab-2, Mouse Monoclonal Antibody.

Marca:  Epredia MS-660-P

Detalles adicionales : Peso : 0.00100kg

 Ver más versiones de este producto

Código de producto. 12641937

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas


Descripción

Descripción

Multi-drug resistance(MDR) associated p-glycoprotein (p170) causes resistance to various drugs in a number of cancer patients. It exists in a soluble and membrane associated form. Soluble P-glycoprotein is detected in extracellular fluids of cancer patients, such as malignant ascites and serum. The Mr of soluble P-glycoprotein is the same as that of membrane bound P-glycoprotein.

Host Species: Mouse

Clone: F4

Isotype: IgG1

Species Reactivity: Human, Chinese Hamster. Does not react with mouse. Others not known.

Epitope: Extracellular amino terminal half. This epitope is resistant to formalin fixation and periodate oxidation and is different from those of other commonly known MAbs to p-glycoprotein, such as C219, JSB-1, and MRK16, which recognize an intracellular and extracellular epitope of p-glycoprotein, respectively.

Immunogen: Human and hamster drug-resistant viable cells followed by crude plasma membranes

Molecular Weight: 170kDa

Positive Control: Drug-resistant cells, Kidney or breast CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Flow Cytometry (Fixed cells only)

Note:

USA: RUO; Int'l: RUO

Especificaciones

Especificaciones

p170/p-Glycoprotein/MDR Ab-2
Monoclonal
200μg/mL
Mouse
1 mL
Cancer and Tumor Biology
Hamster, Human
IgG1
Immunofluorescence, Western Blot
F4
Unconjugated
Human and hamster drug-resistant viable cells followed by crude plasma membranes
RUO
Primary
Concentrated
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales