missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Myoglobin Rabbit Polyclonal Antibody with BSA and azide

Epredia™ Myoglobin, Rabbit Polyclonal Antibody provides accurate, reproducible results.

Marca:  Epredia™ RB-9068-P

Detalles adicionales : Peso : 1.00000kg

 Ver más versiones de este producto

Código de producto. 12683437

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas


Descripción

Descripción

Myoglobin is a cytoplasmic, single chain polypeptide of 153 amino acids and contains a single heme group. Myoglobin is found in skeletal and carciac muscle but not in smooth muscle and functions as an oxygen transporting pigment. Upon damage to the muscle cell due to infarction of a coronary artery, neurological trauma, infection of tumor processes, myoglobin escapes to the environment and can be found in plasma using sensitive assays.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not tested.

Epitope: Not known

Immunogen: Purified human myoglobin

Molecular Weight: 17.8kDa

Positive Control: Skeletal muscle

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Especificaciones

Especificaciones

Myoglobin
Polyclonal
200µg/mL
Rabbit
1 mL
Muscle Biology
Human
IgG
Immunohistochemistry (Paraffin)
Polyclonal
Unconjugated
Purified human myoglobin
RUO
Primary
Concentrated
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales