Learn More
Epredia™ Lab Vision™ MMP-11 (Stromelysin-3) Ab-5 Mouse Monoclonal Antibody, 200μg/mL, biotin labeled with BSA and azide
Ensure accurate, reproducible results in experiments with the Epredia™ MMP-11 (Stromelysin-3) Ab-5, Mouse Monoclonal Antibody.
Marca: Epredia™ MS-1035-P1
Detalles adicionales : Peso : 1.00000kg
Descripción
Stromelysin, a member of the matrix metalloproteinase family, demonstrates wide substrate specificity with the ability to degrade proteoglycan, fibronectin, laminin, casein, and the nonhelical region of collagen. The stromelysin-3 (MMP-11) gene was originally identified on the basis that it is expressed specifically in stromal cells surrounding invasive breast carcinomas. MMP-11 is localized by in situ hybridization to the long arm of chromosome 22. The MMP-11 expression is found more specifically in malignant tumors than in benign ones.Host Species: Mouse
Clone: SL3.05
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Immunogen: Recombinant protein corresponding to active form of human MMP-11
Molecular Weight: ∽60kDa (latent), ∽47kDa (active) and ∽35kDa (degradation product)
Positive Control: Recombinant MMP-11 protein; Placenta
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation (Native verified)
- Affinity Purification (Not verified)
Note:
USA: RUO; Int'l: RUO
Especificaciones
MMP-11 (Stromelysin-3) Ab-5 | |
Monoclonal | |
200μg/mL | |
Mouse | |
500 μL | |
Proteases And Inhibitors | |
Human | |
IgG2a κ |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
SL3.05 | |
Unconjugated | |
Recombinant protein corresponding to active form of human MMP-11 | |
RUO | |
Primary | |
Concentrated |