Learn More
LCK (p56lck) Ab-1 Mouse Monoclonal Antibody, Epredia™
Recommended for Western Blotting, Immunoprecipitation (Denatured verified) and Kinase Assays, Epredia™ LCK (p56lck) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.
Marca: Epredia MS-164-P1ABX
Detalles adicionales : Peso : 1.00000kg
Descripción
p56lck is a membrane-associated non-receptor protein tyrosine kinase from the Src-family of kinases. The phosphorylation status and the activity of p56lck is regulated by the CD45 tyrosine protein phosphatase. p56lck plays a critical role in T cell development and activation. p56lck gene is localized to a site in the genome which undergoes frequent chromosomal abnormalities in lymphomas and neuroblastomas.
Host Species: Mouse
Clone: 3A5
Isotype: IgG2b/κ
Species Reactivity: Human, Mouse and Rat. Others not known.
Epitope: aa 1-225
Immunogen: Recombinant protein corresponding to aa 1-225 of murine p56lck protein
Molecular Weight: 56kDa
Positive Control: Jurkat cells
Cellular Localization: Cell membrane
Recommended for:
- Western Blotting
- Immunoprecipitation (Denatured verified)
- Kinase Assay
Note:
USA: RUO; Int'l: RUO
Especificaciones
LCK (p56superscriptlcksuperscript) Ab-1 | |
Monoclonal | |
1mg/mL | |
Mouse | |
100 μL | |
Cancer and Tumor Biology | |
Human, Mouse, Rat | |
IgG2b κ |
Immunoprecipitation, Western Blot | |
3A5 | |
Unconjugated | |
Recombinant protein corresponding to aa 1-225 of murine p56superscriptlcksuperscript protein | |
RUO | |
Primary | |
Concentrated |