missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Laminin Ab-1 Rabbit Polyclonal Antibody with BSA and azide

Recommended for Immunohistochemistry (Formalin/paraffin). Epredia™ Laminin Ab-1, Rabbit Polyclonal Antibody is one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.

Marca:  Epredia™ RB-082-A1

Detalles adicionales : Peso : 0.00190kg

 Ver más versiones de este producto

Código de producto. 12632777

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos »


Descripción

Descripción

Extracellular matrix (ECM) surrounds and separates most tissue compartments, determines tissue architecture and dynamically interacts with its surrounding cells. The basement membrane is a part of the ECM and is composed of collagens, proteoglycans, elastin and several glycoproteins of which laminin is the most abundant. Laminins are large heterotrimeric, noncollagenous glycoproteins composed of alpha, beta, and gamma chains Dystroglycan is a major laminin receptor in muscle and epithelia, and nidogen is a high-affinity laminin-binding protein important for basement-membrane assembly.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Rat. Others not known.

Epitope: Not determined

Immunogen: Laminin isolated from a rat yolk sac tumor cell line

Positive Control: Skin, Lung, Kidney or Tonsil

Cellular Localization: Basement membrane Ab-1 labels basement membrane surrounding normal vessels, glands, muscles, nerves, fat and deciduas cells separating these from abutting connective tissues.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Especificaciones

Especificaciones

Laminin Ab-1
Polyclonal
Rabbit
Skin, Lung, Kidney or Tonsil
IVD
Primary
Concentrated
Immunohistochemistry (Paraffin)
Unconjugated
Laminin isolated from a rat yolk sac tumor cell line
500 μL
Extracellular Matrix
Human
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales