missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin 19 Ab-4 Mouse Monoclonal Antibody, Epredia™

Obtain accurate, reproducible results in immunohistochemistry and western blotting experiments with  Epredia™ Cytokeratin 19 Ab-4, Mouse Monoclonal Antibody.

Marca:  Epredia MS-1671-P0

Detalles adicionales : Peso : 0.00170kg

 Ver más versiones de este producto

Código de producto. 12600057

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas


Descripción

Descripción

Cytokeratin 19 is a member of type I acidic subfamily of intermediate filaments. It is expressed in various human tissues except in liver. Keratin 19 is not expressed in hepatocytes, therefore, antibody to keratin 19 is useful in the identification of liver metastasis.

Host Species: Mouse

Clone: BA17

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Detergent soluble extract of human mammary epithelium

Molecular Weight: 40 kDa

Positive Control: Colon CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Especificaciones

Especificaciones

Cytokeratin 19 Ab-4
Monoclonal
200μg/mL
Mouse
100 μL
Cytoskeletal Protein
Human
IgG2a κ
Immunohistochemistry (Paraffin), Western Blot
BA17
Unconjugated
Detergent soluble extract of human mammary epithelium
RUO
Primary
Concentrated
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales