Learn More
Epredia™ Lab Vision™ Cytokeratin 18 Rabbit Monoclonal Antibody
Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Cytokeratin 18, Rabbit Monoclonal Antibody.
Marca: Epredia™ RM-2108-S
Detalles adicionales : Peso : 1.00000kg
Descripción
Cytokeratin 18, which belongs to the type A (acidic) subfamily of low molecular weight keratins, exists in combination with cytokeratin 8. Tissues from gastrointestinal tract, respiratory tract and urogenital tract, as well as endocrine and exocrine tissues and mesothelial cells are positive for cytokeratin 18. Typically, it is absent in squamous cell carcinoma.Host Species: Rabbit
Clone: E431-1
Isotype: IgG
Species Reactivity: Human. Others not known.
Immunogen: A synthetic peptide corresponding to residues of the human CK18
Molecular Weight: 45 kDa
Positive Control: Mesothelioma, Pancreas
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Especificaciones
Cytokeratin 18 | |
Monoclonal | |
Unconjugated | |
A synthetic peptide corresponding to residues of the human CK18 | |
IVD | |
Primary | |
Concentrated |
Immunohistochemistry (Paraffin) | |
E431-1 | |
Rabbit | |
1 mL | |
Cancer and Tumor Biology | |
Human | |
IgG |