missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Cytokeratin 18 Rabbit Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Cytokeratin 18, Rabbit Monoclonal Antibody.

Marca:  Epredia™ RM-2108-RQ

Detalles adicionales : Peso : 0.01200kg

 Ver más versiones de este producto

Código de producto. 12683627

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas


Descripción

Descripción

Cytokeratin 18, which belongs to the type A (acidic) subfamily of low molecular weight keratins, exists in combination with cytokeratin 8. Tissues from gastrointestinal tract, respiratory tract and urogenital tract, as well as endocrine and exocrine tissues and mesothelial cells are positive for cytokeratin 18. Typically, it is absent in squamous cell carcinoma.

Host Species: Rabbit

Clone: E431-1

Isotype: IgG

Species Reactivity: Human. Others not known.

Immunogen: A synthetic peptide corresponding to residues of the human CK18

Molecular Weight: 45 kDa

Positive Control: Mesothelioma, Pancreas

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Especificaciones

Especificaciones

Cytokeratin 18
Monoclonal
Unconjugated
A synthetic peptide corresponding to residues of the human CK18
IVD
Primary
Immunohistochemistry (Paraffin)
E431-1
Rabbit
12 mL
Cancer and Tumor Biology
Ready-to-Use
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales