Learn More
Epredia™ Lab Vision™ CEA (Carcinoembryonic Antigen)/CD66e Ab-3 Mouse Monoclonal Antibody, 200μg/mL, biotinylated, BSA and azide
Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence and immunoprecipitation experiments.
Marca: Epredia™ MS-613-R7
Detalles adicionales : Peso : 0.01200kg
Descripción
Host Species: Mouse
Clone: COL-1
Isotype: IgG2a/κ
Species Reactivity: Human. Does not react with rat. Others not known.
Immunogen: Human colon carcinoma extract
Molecular Weight: 180 kDa
Positive Control: LS174T cells, Colon CA
Cellular Localization: Cytoplasmic and luminal membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Native and denatured)
Note:
USA: RUO; Int'l: RUO
Especificaciones
CEA (Carcinoembryonic Antigen)/CD66e Ab-3 | |
Monoclonal | |
Unconjugated | |
7 mL | |
Oncogenesis | |
Human | |
IgG2a κ |
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
COL-1 | |
Mouse | |
RUO | |
Primary | |
Prediluted |