Learn More
c-myc Ab-2 Mouse Monoclonal Antibody, Epredia™
Epredia™ c-myc Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.
Marca: Epredia MS-139-P1ABX
Detalles adicionales : Peso : 1.00000kg
Descripción
c-myc is involved in the control of cell proliferation and differentiation and is amplified and/or overexpressed in a variety of tumors.Host Species: Mouse
Clone: 9E10.3
Isotype: IgG1/κ
Species Reactivity: Human. Does not react with mouse, rat and chicken. Others not tested.
Epitope: aa 410-419
Immunogen: A synthetic peptide, aa 408-439, (AEEQKLISEEDLLRKRREQLKHKLEQLRNSCA) from C-terminus of human c-myc
Molecular Weight: 64-67 kDa
Positive Control: Raji cells, Breast CA
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Flow Cytometry
- Electron Microscopy
- Affinity Purification, Immunoprecipitation and Western Blotting of fusion proteins labeled with EQKLISEEDL tag
Note:
USA: RUO; Int'l: RUO
Especificaciones
c-myc Ab-2 | |
Monoclonal | |
1mg/mL | |
Mouse | |
100 μL | |
Transcription Regulation | |
Human | |
IgG2a κ |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
9E10.3 | |
Unconjugated | |
A synthetic peptide, aa 408-439, (AEEQKLISEEDLLRKRREQLKHKLEQLRNSCA) from C-terminus of human c-myc | |
RUO | |
Primary | |
Concentrated |