missing translation for 'onlineSavingsMsg'
Learn More

Amyloid Precursor Protein (APP) Rabbit Polyclonal Antibody, Epredia™

Obtain accurate, reproducible results in immunohistochemistry and western blotting experiments.

Marca:  Epredia RB-9023-P1

Detalles adicionales : Peso : 1.00000kg

 Ver más versiones de este producto

Código de producto. 12633337

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas


Descripción

Descripción

Recognizes APP (b-APP770), MW 110kDa. Predicted to recognize Β-APP695 and Β-APP751. Amyloid precursor protein (APP) and APP-like proteins are transmembrane glycoproteins with a similar modular domain structure.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not tested.

Epitope: N-terminal

Immunogen: A synthetic peptide corresponding to N-terminal of human APP

Molecular Weight: 110kDa

Positive Control: Brain

Cellular Localization: Cytoplasmic and extracellular

This antibody is excellent for staining of formalin-fixed, paraffin-embedded tissues

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
Especificaciones

Especificaciones

Amyloid Precursor Protein (APP)
Polyclonal
Rabbit
500 μL
Neuroscience
Human, Mouse, Rat
IgG
Western Blot
Unconjugated
A synthetic peptide corresponding to N-terminal of human APP
IVD
Primary
Concentrated
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales