Learn More
Actin, Smooth Muscle, Ab-1 Mouse Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry and immunofluorescence experiments with Epredia™ Actin, Smooth Muscle Ab-1, Mouse Monoclonal Antibody.
Marca: Epredia MS-113-P1ABX
Detalles adicionales : Peso : 1.00000kg
Descripción
Host Species: Mouse
Clone: 1A4; also known as asm-1
Isotype: IgG2a/κ
Species Reactivity: Human, Baboon, Cow, Rabbit, Mouse, Rat and Chicken. Others not known.
Epitope: Acetyl group and the first four amino acids on the N-terminal end of the peptidic chain of α-smooth actin
Immunogen: N-terminal decapeptide of α-smooth muscle isoform of actin; acetylated at the N-terminus
Positive Control: Smooth muscle or leiomyosarcoma
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Immunofluorescence
Especificaciones
Actin, Smooth Muscle Ab-1 | |
Monoclonal | |
1mg/mL | |
Mouse | |
100 μL | |
Muscle Biology | |
Bovine, Chicken, Human, Mouse, Non-human Primate, Rabbit, Rat | |
IgG2a κ |
Immunofluorescence, Immunohistochemistry (Paraffin) | |
1A4 ; also known as asm-1 | |
Unconjugated | |
N-terminal decapeptide of alpha-smooth muscle isoform of actin; acetylated at the N-terminus | |
IVD | |
Primary | |
Concentrated |