Learn More
Clorhidrato de L-cisteína, monohidrato, multicompendial, U.S.P., J.T.Baker™
Catalog Number p-5391627
Click to view available options
Cantidad:
100 g
1 kg
Envase:
Frasco de polietileno
7048-04-6
C3H10ClNO3S
175.63
QIJRTFXNRTXDIP-JIZZDEOASA-N
l-cysteine hydrochloride monohydrate, h-cys-oh.hcl.h2o, l-cysteine hydrochloride hydrate, l-cysteine monohydrate monochloride, r-2-amino-3-mercaptopropanoic acid hydrochloride hydrate, unii-zt934n0x4w, l-cysteine hydrate hydrochloride, cysteine hydrochloride monohydrate, l, h-cys-ohhclh2o, cysteine hcl
23462
Ácido (2R)-2-amino-3-sulfanilpropanoico;hidrato;clorhidrato
C(C(C(=O)O)N)S.O.Cl
7048-04-6
C3H10ClNO3S
175.63
QIJRTFXNRTXDIP-JIZZDEOASA-N
l-cysteine hydrochloride monohydrate, h-cys-oh.hcl.h2o, l-cysteine hydrochloride hydrate, l-cysteine monohydrate monochloride, r-2-amino-3-mercaptopropanoic acid hydrochloride hydrate, unii-zt934n0x4w, l-cysteine hydrate hydrochloride, cysteine hydrochloride monohydrate, l, h-cys-ohhclh2o, cysteine hcl
23462
Ácido (2R)-2-amino-3-sulfanilpropanoico;hidrato;clorhidrato
C(C(C(=O)O)N)S.O.Cl
Nombre del producto químico o material L-Cysteine Hydrochloride
Nombre de nota Monohydrate
Envase Frasco de polietileno
Formula Weight (peso de la fórmula) 175.64
Grado Multi-Compendial/Farmacopea Europea
Cantidad 100 g
Gravedad específica 1L = 1.50kg