Learn More
KRT71 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA528558
Detalles adicionales : Peso : 0.01000kg
Descripción
Recommended positive controls: HepG2. Predicted reactivity: Mouse (91%), Rat (91%), Dog (91%), Cat (93%), Sheep (91%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
KRT71 Polyclonal antibody specifically detects KRT71 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Especificaciones
KRT71 | |
Polyclonal | |
Unconjugated | |
KRT71 | |
CK-71; cytokeratin-71; hK6irs; hK6irs1; HYPT13; K6IRS1; K71; KB34; Keratin 6 irs; keratin 71; keratin 71, type II; keratin, type II cytoskeletal 71; Keratin-71; KRT6IRS; KRT6IRS1; KRT71; Type II inner root sheath-specific keratin-K6irs1; type-II keratin Kb34 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
112802 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.91 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q3SY84 | |
KRT71 | |
Recombinant fragment corresponding to a region within amino acids 211 and 523 of Human KRT71 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |