missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLHL10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 25591-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Especificaciones
KLHL10 | |
Polyclonal | |
Unconjugated | |
KLHL10 | |
kelch like 10 (Drosophila), Kelch like protein 10, KLHL10 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
303533, 317719, 66720 | |
-20°C | |
Liquid |
Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6JEL2, Q6JEL3, Q9D5V2 | |
KLHL10 | |
KLHL10 Fusion Protein Ag22368 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |