Learn More
Kinesin 5A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA529820
Detalles adicionales : Peso : 0.01000kg
Descripción
Recommended positive controls: HeLa, mouse brain, rat brain. Predicted reactivity: Mouse (95%), Rat (96%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the kinesin family of proteins. Members of this family are part of a multisubunit complex that functions as a microtubule motor in intracellular organelle transport. Mutations in this gene cause autosomal dominant spastic paraplegia 10.Especificaciones
Kinesin 5A | |
Polyclonal | |
Unconjugated | |
KIF5A | |
D10Bwg0738e; D12S1889; Khc; Kiaa4086; Kif5; Kif5a; kif5a {ECO:0000312; KIF5A variant protein; kinesin family member 5A; kinesin heavy chain isoform 5A; kinesin heavy chain isoform 5A; kinesin heavy chain; kinesin heavy chain neuron-specific 1; kinesin, heavy chain, neuron-specific; Kns; mKIAA4086; MY050; Neuronal kinesin heavy chain; NKHC; Nkhc1; RGD:1303035}; SPG10 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16572, 314906, 3798 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.23 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
P33175, Q12840, Q6QLM7 | |
KIF5A | |
Recombinant fragment corresponding to a region within amino acids 508 and 800 of Human Kinesin 5A | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |