missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIF13A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 27210-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a member of the kinesin family of microtubule-based motor proteins that function in the positioning of endosomes. This family member can direct mannose-6-phosphate receptor-containing vesicles from the trans-Golgi network to the plasma membrane, and it is necessary for the steady-state distribution of late endosomes/lysosomes. It is also required for the translocation of FYVE-CENT and TTC19 from the centrosome to the midbody during cytokinesis, and it plays a role in melanosome maturation. Alternative splicing of this gene results in multiple transcript variants.Especificaciones
KIF13A | |
Polyclonal | |
Unconjugated | |
KIF13A | |
KIF13A, kinesin family member 13A, Kinesin like protein KIF13A, Kinesin like protein RBKIN, RBKIN | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
63971 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9H1H9 | |
KIF13A | |
KIF13A Fusion Protein Ag24356 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |