Learn More
KCTD15 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA530810
Detalles adicionales : Peso : 0.01000kg
Descripción
Recommended positive controls: NT2D1, IMR32. Predicted reactivity: Mouse (98%), Rat (98%), Xenopus laevis (88%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
KCTD15 Polyclonal antibody specifically detects KCTD15 in Human samples. It is validated for Immunocytochemistry, Western Blot.Especificaciones
KCTD15 | |
Polyclonal | |
Unconjugated | |
KCTD15 | |
BC031749; BTB/POZ domain-containing protein KCTD15; Kctd15; potassium channel tetramerisation domain containing 15; potassium channel tetramerization domain containing 15; potassium channel tetramerization domain-containing protein 15 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79047 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.89 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q96SI1 | |
KCTD15 | |
Recombinant protein fragment corresponding to a region within amino acids 24 and 223 of Human KCTD15 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |