missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNK5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14136-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
KCNK5 is one of the members of the superfamily of potassium channel proteins containing two pore-forming P domains. KCNK5 is mainly expressed in the cortical distal tubules and collecting ducts of the kidney. The protein is highly sensitive to external pH and this, in combination with its expression pattern, suggests it may play an important role in renal potassium transport.Especificaciones
KCNK5 | |
Polyclonal | |
Unconjugated | |
KCNK5 | |
K2p5.1, KCNK5, TASK 2, TASK2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16529, 364241, 8645 | |
-20°C | |
Liquid |
Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O95279 | |
Kcnk5 | |
KCNK5 Fusion Protein Ag5355 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |