Learn More
Kanamycin Sulfate, Ultrapure, Thermo Scientific Chemicals
40.25€ - 174.00€
Identificadores de productos químicos
CAS | 25389-94-0 |
---|---|
Fórmula molecular | C18H38N4O15S |
Peso molecular (g/mol) | 582.575 |
Número MDL | MFCD00070253 |
Clave InChI | OOYGSFOGFJDDHP-IZQIRFRQSA-N |
Sinónimo | Kanamycin A |
PubChem CID | 134129479 |
Nombre IUPAC | (2 R, 3 S, 4 S, 5 S, 6 R)-2-(aminometil)-6-[(1 R, 2 S, 3 S, 4 R, 6 S)-4,6-diamino-3-[(2 S, 3 R, 4 S, 5 R, 6 R)-4-amino-3,5-dihidroxi-6-(hidroximetil)oxan-2-il]oxi-2-hidroxiciclohexil]oxioxano-3,4,5-triol; ácido sulfúrico |
SMILES | C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O |
Descripción
A selectable marker for cosmid vectors used in transfection of mammalian cells.
Mode of Action: Binds to ribosomal components and inhibits protein synthesis. Kanamycin is inactivated by an aminoglycoside phospho transferase (MW 25,000) in the periplasmic space.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Selectable marker for cosmid vectors used in transfection of mammalian cells
- Mode of Action: Binds to ribosomal components and inhibits protein synthesis
- Kanamycin is inactivated by aminoglycoside phospho transferase (MW 25,000) in periplasmic space
Identificadores de productos químicos
25389-94-0 | |
582.575 | |
OOYGSFOGFJDDHP-IZQIRFRQSA-N | |
134129479 | |
C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O |
C18H38N4O15S | |
MFCD00070253 | |
Kanamycin A | |
(2 R, 3 S, 4 S, 5 S, 6 R)-2-(aminometil)-6-[(1 R, 2 S, 3 S, 4 R, 6 S)-4,6-diamino-3-[(2 S, 3 R, 4 S, 5 R, 6 R)-4-amino-3,5-dihidroxi-6-(hidroximetil)oxan-2-il]oxi-2-hidroxiciclohexil]oxioxano-3,4,5-triol; ácido sulfúrico |
Especificaciones
Kanamycin sulfate | |
Polvo | |
C18H38N4O15S | |
Kanamycin A | |
C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O | |
582.575 | |
582.58 |
Kanamycin Sulfate | |
25389-94-0 | |
MFCD00070253 | |
OOYGSFOGFJDDHP-IZQIRFRQSA-N | |
(2 R, 3 S, 4 S, 5 S, 6 R)-2-(aminometil)-6-[(1 R, 2 S, 3 S, 4 R, 6 S)-4,6-diamino-3-[(2 S, 3 R, 4 S, 5 R, 6 R)-4-amino-3,5-dihidroxi-6-(hidroximetil)oxan-2-il]oxi-2-hidroxiciclohexil]oxioxano-3,4,5-triol; ácido sulfúrico | |
134129479 |
Seguridad y manipulación
P201-P202-P260-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c
H317-H334-H360FD
missing translation for 'einecsNumber' : 246-933-9
missing translation for 'rtecsNumber' : NZ3225030
missing translation for 'tsca' : No
Recommended Storage : Mantener en frío
RUO – Research Use Only