Learn More
STEMCELL Technologies ITS Supplement-A
For neural and pancreatic differentiation of mouse ES and iPS cells
Marca:  STEMCELL Technologies 07151
Detalles adicionales : Peso : 0.01330kg
Código de producto. 17109241
  • 405.00€ / 5 ml
Fecha estimada de envío: 20-05-2024
para ver el stock
Descripción
Descripción
  • ITS Supplement-A is a medium supplement containing insulin, holo-transferrin (iron-saturated) and selenium in phosphate-buffered saline
  • It is recommended for use in the in vitro differentiation of mouse embryonic stem (ES) cells and mouse induced pluripotent stem (iPS) cells to neural and pancreatic-like cell types
Especificaciones
Especificaciones
Pluripotent Stem Cells
Cell Culture, Differentiation
5 mL