Learn More
STEMCELL Technologies ITS Supplement-A
For neural and pancreatic differentiation of mouse ES and iPS cells
Marca: STEMCELL Technologies 07151
Detalles adicionales : Peso : 0.01330kg
Descripción
- ITS Supplement-A is a medium supplement containing insulin, holo-transferrin (iron-saturated) and selenium in phosphate-buffered saline
- It is recommended for use in the in vitro differentiation of mouse embryonic stem (ES) cells and mouse induced pluripotent stem (iPS) cells to neural and pancreatic-like cell types
Especificaciones
Pluripotent Stem Cells | |
Cell Culture, Differentiation | |
5 mL |