Learn More
ITPRIPL2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA565323
Detalles adicionales : Peso : 0.01000kg
Descripción
Immunogen sequence: WLRDCKPFADA FCVDVRGRRH LSATLVLRWF QSHLQRSLAT VRYSLEGRCR VTLTPGGLEQ PPTLHILPCR TDYGCCRLSM AVRLIPAVH Highest antigen sequence identity to the following orthologs - mouse 92%, rat 24%.
ITPRIPL2 is a protein coding gene.Especificaciones
ITPRIPL2 | |
Polyclonal | |
Unconjugated | |
ITPRIPL2 | |
inositol 1,4,5-triphosphate receptor interacting protein-like 2; inositol 1,4,5-triphosphate receptor-interacting protein-like 2; inositol 1,4,5-trisphosphate receptor interacting protein-like 2; inositol 1,4,5-trisphosphate receptor-interacting protein-like 2; ITPRIPL2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
162073 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q3MIP1 | |
ITPRIPL2 | |
Recombinant Human ITPRIPL2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only