missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(Iii) Nitrate Nonahydrate, Honeywell Fluka™
Click to view available options
Cantidad:
100 g
250 g
500 g
1 kg
2.5 kg
Descripción
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Especificaciones
Nombre del producto químico o material | Iron(Iii) Nitrate Nonahydrate |
Cantidad | 2.5 kg |
Fórmula lineal | Fe(NO3)3 · 9H2O |
Número UN | 3260 |
SMILES | [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Fe+3] |
Formula Weight (peso de la fórmula) | 404g/mol |
Porcentaje de pureza | ≥98% |
Grado | ACS Reagent |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'