Learn More
IRF5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA517255
Detalles adicionales : Peso : 0.01000kg
Descripción
It is not recommended to aliquot this antibody.
This gene encodes a member of the interferon regulatory factor (IRF) family, a group of transcription factors with diverse roles, including virus-mediated activation of interferon, and modulation of cell growth, differentiation, apoptosis, and immune system activity. Members of the IRF family are characterized by a conserved N-terminal DNA-binding domain containing tryptophan (W) repeats. Alternative splice variants encoding different isoforms exist.Especificaciones
IRF5 | |
Polyclonal | |
Unconjugated | |
IRF5 | |
AW491843; Interferon regulatory factor 5; interferon regulatory factor 5 bone marrow; IRF5; IRF-5; mirf5; OTTHUMP00000212907; SLEB10 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27056, 296953, 3663 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
40 μg/mL | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
P56477, Q13568 | |
IRF5 | |
Synthetic peptide corresponding to residues surrounding Ser172 of mouse IRF-5 protein | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |