Learn More
IRAK1 293T Cell Overexpression Lysate (Denatured), Abnova
Denatured IRAK1 293 cell overexpression lysate
Marca: Abnova H00003654-T01.100uL
Detalles adicionales : Peso : 0.00010kg
Beschreibung
This gene encodes the interleukin-1 receptor-associated kinase 1, one of two putative serine/threonine kinases that become associated with the interleukin-1 receptor (IL1R) upon stimulation. This gene is partially responsible for IL1-induced upregulation of the transcription factor NF-kappa B. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. (provided by RefSeq)
- Transfected cell line: 293T
- Plasmid: pCMV-IRAK1 full-length
- Host: Human
- Molecular weight: 69.74kDa
- Storage Buffer:1X Sample Buffer (50mM Tris-HCl, 2% SDS, 10% glycerol, 300mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blotting
Spezifikation
Western Blot | |
IRAK1 293T Cell Transient Overexpression Lysate(Denatured) | |
69.74 | |
pCMV-IRAK1 full-length | |
100 μL | |
Human |
293T | |
BC014963.1 | |
6.8 | |
Transient overexpression cell Lysate was tested with Anti-IRAK1 antibody (H00003654-B01) by western blotting | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Sicherheit und Handhabung
- 100UL IRAK1 293T Cell Transient Overexpression Lys
Signalwort
- Atención
Gefahrenkategorie
- Sensibilización cutánea Categoría 1
Gefahrenhinweis
- H317-Puede provocar una reacción alérgica en la piel.
Sicherheitshinweis
- P261-Evitar respirar el polvo/el humo/el gas/la niebla/los vapores/el aerosol.
- P272-Las prendas de trabajo contaminadas no podrán sacarse del lugar de trabajo.
- P280-Llevar guantes/prendas/gafas/máscara de protección.
Ergänzende Informationen
- MIXTURE LIST-Contém: Mercaptoethanol