Learn More
Invitrogen™ TOPO™ TA Cloning™ Kit, Dual Promoter, with One Shot™ TOP 10F' Chemically Competent E.coli, no manual
Designed for cloning PCR products directly from a PCR reaction in just 5 minutes
Marca: Invitrogen™ KNM465001
Detalles adicionales : Peso : 0.00500kg
Includes: Topoisomerase I-activated pCR™II-TOPO™ vector; PCR buffer; Salt solution; dNTPs; Control template; M13 forward and reverse primers; Control PCR primers; Sterile water; One Shot™ Chemically Competent or Electrocomp™ E. coli; S.O.C. medium; Supercoiled pUC19 control plasmid.
Descripción
TOPO TA Cloning kits• Alta eficiencia
• 'One-step'
Especificaciones
Kit, Cloning | |
TOP10F´ | |
25 Reactions |
Box 1: Topoisomerase I-activated pCRII-TOPO vector, PCR buffer, Salt solution, dNTPs, Control template, M13 forward and reverse primers, Control PCR primers, Sterile water. Box 2: One Shot Chemically Competent or Electrocomp E. coli, S.O.C. medium, Supercoiled pUC19 control plasmid | |
Cloning, PCR Cloning |
For Research Use Only. Not for use in diagnostic procedures.