missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta-1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 26918-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Integrins are heterodimeric proteins made up of alpha and beta subunits. At least 18 alpha and 8 beta subunits have been described in mammals. Integrin family members are membrane receptors involved in cell adhesion and recognition in a variety of processes including embryogenesis, hemostasis, tissue repair, immune response and metastatic diffusion of tumor cells. This gene encodes a beta subunit. Multiple alternatively spliced transcript variants which encode different protein isoforms have been found for this gene.Especificaciones
Integrin beta-1 | |
Polyclonal | |
Unconjugated | |
ITGB1 | |
CD29, FNRB, GPIIA, Integrin beta 1, Integrin beta-1, ITGB1, MDF2, MSK12, VLA 4 subunit beta, VLA BETA, VLAB | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3688 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P05556 | |
ITGB1 | |
Integrin beta-1 Fusion Protein Ag25426 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |