missing translation for 'onlineSavingsMsg'
Learn More

Insulin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 15848-1-AP-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16815224

  • 140.00€ / 20 microlitros
Fecha estimada de envío: 21-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Insulin is a peptide hormone secreted by beta cells of the pancreatic islets. It regulates carbohydrate, protein and lipid metabolism by enhancing membrane transport of glucose, amino acids, and certain ions. It also promotes glycogen storage, formation of triglycerides and synthesis of proteins and nucleic acids. Insulin is 51-amino acid polypeptide product produced from a precursor peptide, proinsulin. Proinsulin is post-translationally cleaved into two chains (peptide A and peptide B) that are covalently linked via two disulfide bonds and one molecule of C-peptide. Insulin deficiency results in diabetes mellitus, one of the leading causes of morbidity and mortality in the general population. Insulin is also present in tumors of b-cell origin such as insulinoma.
Especificaciones

Especificaciones

Insulin
Polyclonal
Unconjugated
INS
ILPR, INS, insulin, IRDN
Rabbit
Antigen Affinity Chromatography
RUO
3630
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence
0.28 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P01308
INS
Insulin Fusion Protein Ag8630
20 μL
Primary
Human, Rat, Mouse
Antibody
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales