Learn More
Fisherbrand™ Inkless Chart Recorders for Isotemp Ultra-Low Temperature Chest Freezers
Record seven days of temperature performance at temperatures to -150°C using Fisherbrand™ Isotemp Chart Recorders for Ultra-Low Temperature Freezers.
Brand: Fisherbrand™
Additional Details : Peso : 0.40000kg
Description
- Registrador de siete días
- Funciona a hasta -150 °C
- Instalado en fábrica
Specifications
Factory Installed | |
-150°C | |
7 Days |
Chart Recorder | |
Isotemp 13 and 20 cu. ft. Chest Ultra-Low Temperature Freezer, Ultracongelador horizontal Isotemp de 368 y 566 l (13 y 20 pies cúbicos) |