Learn More
Gasa Inconel™ 600, malla 20 tejida a partir de alambre de 0,41 mm de diámetro, Thermo Scientific™
Marca: Thermo Scientific Alfa Aesar 046510.HB
Detalles adicionales : Peso : 0.01000kg
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Especificaciones
Inconel™ 600 gauze | |
Ni:Cr:Fe; 77:15.5:7.5 wt% | |
300 x 300 mm | |
Tela de alambre |
20 mesh woven from 0.41mm (0.016 in.) dia. wire | |
MFCD01741927 | |
20 Mesh |