missing translation for 'onlineSavingsMsg'
Learn More

IL36RN Mouse anti-Human, Clone: 5F11G6, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 60298-1-IG-150UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16877375

  • 399.00€ / 150 microlitros
Fecha estimada de envío: 24-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The protein encoded by this gene is a member of the interleukin 1 cytokine family. This cytokine was shown to specifically inhibit the activation of NF-kappaB induced by interleukin 1 family, member 6 (IL1F6). This gene and eight other interleukin 1 family genes form a cytokine gene cluster on chromosome 2. Two alternatively spliced transcript variants encoding the same protein have been reported.
Especificaciones

Especificaciones

IL36RN
Monoclonal
1.37 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9UBH0
IL36RN
IL36RN Fusion Protein Ag3780
150 μL
Primary
Human
Antibody
IgG2b
Western Blot
5F11G6
Unconjugated
IL36RN
FIL1, FIL1 delta, FIL1(DELTA), FIL1D, IL 1 delta, IL 1 related protein 3, IL 1F5, IL 1HY1, IL 1L1, IL 1ra homolog 1, IL 1RP3, IL1F5, IL1HY1, IL1L1, IL1RP3, Interleukin 1 delta, Interleukin 1 family member 5, Interleukin 1 HY1, Interleukin 1 like protein 1
Mouse
Protein A
RUO
26525
-20°C
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales