missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL10RB Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15102-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Natural killer (NK) cells in teleosts and their evolutionary homologue are a subpopulation of lymphocytes with properties that distinguish them from either B- or T-cells. NK cells are important effectors of innate immunity where they release cytokines, which in turn up-regulate other immunological functions. Monoclonal antibodies have been used to identify different surface antigens present on NK cells. These surface antigens have not only been used to identify NK cells, but also their functionally distinct subsets. The Cluster of Differentiation (CD) nomenclature was established to standardize the naming of NK cells.Especificaciones
IL10RB | |
Polyclonal | |
Unconjugated | |
IL10RB | |
CDW210B, CRF2 4, CRFB4, D21S58, D21S66, IL 10 receptor subunit beta, IL 10R subunit 2, IL 10R subunit beta, IL 10R2, IL 10RB, IL10RB, interleukin 10 receptor, beta | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16155, 3588 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q08334, Q61190 | |
IL10RB | |
IL10RB Fusion Protein Ag7445 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |