Learn More
IgG1 Rat, PE, Clone: RTK2071, Isotype Control, STEMCELL Technologies
Rat Monoclonal Antibody
Marca:  Stemcell Technologies 60075PE
Detalles adicionales : Peso : 0.00110kg
 Ver más versiones de este producto
Código de producto. 17188891
  • 140.98€ / 100 microgramos
Fecha estimada de envío: 10-11-2022
para ver el stock
Añadir a la cesta
Descripción
Descripción
  • The RTK2071 antibody (IgG1, kappa) is suitable for use as an isotype-matched control antibody in several applications to estimate the degree of non-specific binding by an antigen-specific antibody
  • Ideally, the isotype control should have the same subclass of heavy chain (IgA, IgD, IgE, IgG, or IgM) and light chain (kappa or lambda) as the specific antibody being used
  • If a conjugated antibody is used, an isotype control conjugated to the same molecule (e.g. fluorochrome) should be chosen
  • The use of an appropriate isotype control helps confirm the specificity of the antigen-specific antibody and indicates non-specific binding that may result from binding to Fc receptors or other cell components
  • This antibody clone has been verified for use as an isotype control antibody for assessing non-specific binding to cells in applications such as flow cytometry and immunofluorescence microscopy applications (surface and intracellular staining)
Especificaciones
Especificaciones
IgG1
Monoclonal
PE
100 μg
IgG1 κ
Immunofluorescence, Immunocytochemistry, Flow Cytometry, Western Blot, Immunohistochemistry
RTK2071
Rat
Isotype Control
Other