Learn More
IgG Rabbit, Clone: IG1707R, enQuireBio™
Rabbit Monoclonal Antibody
Marca: EnquireBio 3500-RBM9-P0
Detalles adicionales : Peso : 0.01000kg
Especificaciones
IgG | |
Cytoplasm, Cell Surface and Secreted | |
IG1707R | |
Unconjugated | |
IGHG | |
Purified human Ig Gamma Chain | |
293T, Raji or hPBL cells. Tonsil or Spleen. | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with or without 0.05% BSA and 0.05% azide. | |
Rabbit | |
75kDa | |
Protein A/G Purified | |
RUO | |
3500 | |
IgG κ |