Learn More
IFN gamma Rat, PE, Clone: XMG1.2, enQuireBio™
Rat Monoclonal Antibody
Marca: EnquireBio QAB83-PE-25ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
IFN gamma | |
Monoclonal | |
0.5mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2 | |
Ifng | |
Protein A/G Purified | |
RUO | |
15978 | |
IgG1 κ |
Flow Cytometry | |
XMG1.2 | |
PE | |
NP_032363 | |
Rat | |
25 μg | |
Primary | |
Store at 2-4°C. |