missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN Gamma Mouse anti-Human, CoraLite 647, Clone: 1E10G7, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech CL647-69007-100UL
Detalles adicionales : Peso : 0.00010kg
Descripción
IFN Gamma Monoclonal antibody specifically detects IFN Gamma in Human samples. It is validated for FC (Intra)Especificaciones
IFN Gamma | |
Monoclonal | |
1000 μg/mL | |
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
Mouse | |
100 μL | |
3458 | |
Store at -20°C | |
Liquid |
Flow Cytometry | |
1E10G7 | |
CoraLite 647 | |
BC000627 | |
Protein G purification | |
Primary | |
Human | |
Antibody | |
IgG1 |