Learn More
Iduronate 2 Sulfatase Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA545170
Detalles adicionales : Peso : 0.00010kg
Descripción
Peptide sequence: SFPPYHPSSE KYENTKTCRG PDGELHANLL CPVDVLDVPE GTLPDKQSTE Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%
Iduronate 2 Sulfatase Polyclonal antibody specifically detects Iduronate 2 Sulfatase in Human samples. It is validated for Western Blot.Especificaciones
Iduronate 2 Sulfatase | |
Polyclonal | |
Unconjugated | |
Ids | |
alpha-L-iduronate sulfate sulfatase; AW214631; Ids; Iduronate 2-sulfatase; iduronate 2-sulfatase (Hunter syndrome); iduronate 2-sulfatase 14 kDa chain; Iduronate 2-sulfatase 42 kDa chain; iduronate sulfatase; iduronate-2-sulfatase; idursulfase; MPS2; SIDS | |
Rabbit | |
Affinity Chromatography | |
RUO | |
3423 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P22304 | |
Ids | |
Synthetic peptide directed towards the N-terminal of human IDS | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |