missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plas Labs™ Hypalon Gloves
Click to view available options
Tamaño:
8
Descripción
- White one-piece gauntlet style standard gloves suitable for use with most Plas-Labs™ gloveboxes
- Constructed from Hypalon™ chlorosulfonated polyethylene (CSPE) synthetic rubber
- Resistant to abrasion, oxidising chemicals ozone and UV light
- Ambidextrous fit
- Sold in pairs
- Range of hand sizes and port diameters
Especificaciones
Material | CSM |
Tipo de producto | Gloves |
Tamaño | 8 |
Grosor (métrico) | 0.15 mm |
Estilo de puño de los guantes | 8 in. diameter |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'