missing translation for 'onlineSavingsMsg'
Learn More

Hydroxylamine Hydrochloride, ReagentPlus™, Honeywell

Catalog Number p-7071830
Click to view available options
Cantidad:
100 g
500 g
1 kg
3 kg
Este artículo no se puede devolver. Vea la política de devoluciones
5470-11-1
ClH4NO
69.49
MFCD00051089
WTDHULULXKLSOZ-UHFFFAOYSA-N
hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci
443297
CHEBI:5807
hydroxylamine hydrochloride
Cl.NO
Este artículo no se puede devolver. Vea la política de devoluciones
5470-11-1
ClH4NO
69.49
MFCD00051089
WTDHULULXKLSOZ-UHFFFAOYSA-N
hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci
443297
CHEBI:5807
hydroxylamine hydrochloride
Cl.NO
Nombre del producto químico o material Hydroxylamine Hydrochloride
Cantidad 100 g
Fórmula lineal NH2OH · HCl
Número UN 2923
Beilstein 3539763
Formula Weight (peso de la fórmula) 69.49g/mol
Porcentaje de pureza 99%
Grado ReagentPlus™
Densidad 1.67g/mL at 25°C
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'