missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ZBTB3 Synthetic Peptide
Synthetic Peptide
Marca: Invitrogen™ PEP1197
Detalles adicionales : Peso : 0.01000kg
Descripción
ZBTB3 Synthetic Peptide, PEP-1197, from Invitrogen.
This peptide corresponds to 17 amino acid peptide near the carboxy terminus of human ZBTB3.PEP-1197 can be used as a blocking peptide with polyclonal antibody PA5-21083.Especificaciones
79842 | |
purified | |
Human ZBTB3 | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
Q9H5J0 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
ZBTB3 | |
0.02% sodium azide | |
50 μg | |
PBS |