missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human UBE2E1 Full-length ORF (NP_003332.1, 1 a.a. - 193 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00007324-P01.10ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene. [provided by RefSeq]
Sequence: MSDDDSRASTSSSSSSSSNQQTEKETNTPKKKESKVSMSKNSKLLSTSAKRIQKELADITLDPPPNCSAGPKGDNIYEWRSTILGPPGSVYEGGVFFLDITFTPEYPFKPPKVTFRTRIYHCNINSQGVICLDILKDNWSPALTISKVLLSICSLLTDCNPADPLVGSIATQYMTNRAEHDRMARQWTKRYATEspecificaciones
NP_003332.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
47.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
UBCH6 | |
UBE2E1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
7324 | |
UBE2E1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSDDDSRASTSSSSSSSSNQQTEKETNTPKKKESKVSMSKNSKLLSTSAKRIQKELADITLDPPPNCSAGPKGDNIYEWRSTILGPPGSVYEGGVFFLDITFTPEYPFKPPKVTFRTRIYHCNINSQGVICLDILKDNWSPALTISKVLLSICSLLTDCNPADPLVGSIATQYMTNRAEHDRMARQWTKRYAT | |
RUO | |
UBE2E1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |