missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human TAPBPL Full-length ORF (NP_060479.2, 1 a.a. - 468 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00055080-P01.10ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Tapasin, or TAPBP (MIM 601962), is a member of the variable-constant Ig superfamily that links major histocompatibility complex (MHC) class I molecules to the transporter associated with antigen processing (TAP; see MIM 170260) in the endoplasmic reticulum (ER). The TAPBP gene is located near the MHC complex on chromosome 6p21.3. TAPBPL is a member of the Ig superfamily that is localized on chromosome 12p13.3, a region somewhat paralogous to the MHC.[supplied by OMIM]
Sequence: MGTQEGWCLLLCLALSGAAETKPHPAEGQWRAVDVVLDCFLVKDGAHRGALASSEDRARASLVLKQVPVLDDGSLEDFTDFQGGTLAQDDPPIIFEASVDLVQIPQAEALLHADCSGKEVTCEISRYFLQMTETTVKTAAWFMANVQVSGGGPSISLVMKTPRVAKNEVLWHPTLNLPLSPQGTVRTAVEFQVMTQTQSLSFLLGSSASLDCGFSMAPGLDLISVEWRLQHKGRGQLVYSWTAGQGQAVRKGATLEPAQLGMARDASLTLPGLTIQDEGTYICQITTSLYRAQQIIQLNIQASPKVRLSLANEALLPTLICDIAGYYPLDVVVTWTREELGGSPAQVSGASFSSLRQSVAGTYSISSSLTAEPGSAGATYTCQVTHISLEEPLGASTQVVPPERRTALGVIFASSLFLLALMFLGLQRRQAPTGLGLLQAERWETTSCADTQSSHLHEDRTARVSQPSEspecificaciones
NP_060479.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
76.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ10143/TAPBP-R/TAPBPR | |
TAPBPL | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
55080 | |
TAPBPL (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGTQEGWCLLLCLALSGAAETKPHPAEGQWRAVDVVLDCFLVKDGAHRGALASSEDRARASLVLKQVPVLDDGSLEDFTDFQGGTLAQDDPPIIFEASVDLVQIPQAEALLHADCSGKEVTCEISRYFLQMTETTVKTAAWFMANVQVSGGGPSISLVMKTPRVAKNEVLWHPTLNLPLSPQGTVRTAVEFQVMTQTQSLSFLLGSSASLDCGFSMAPGLDLISVEWRLQHKGRGQLVYSWTAGQGQAVRKGATLEPAQLGMARDASLTLPGLTIQDEGTYICQITTSLYRAQQIIQLNIQASPKVRLSLANEALLPTLICDIAGYYPLDVVVTWTREELGGSPAQVSGASFSSLRQSVAGTYSISSSLTAEPGSAGATYTCQVTHISLEEPLGASTQVVPPERRTALGVIFASSLFLLALMFLGLQRRQAPTGLGLLQAERWETTSCADTQSSHLHEDRTARVSQPS | |
RUO | |
TAPBPL | |
Wheat Germ (in vitro) | |
GST | |
Liquid |