Learn More
Abnova™ Human SYNE1 Partial ORF (NP_892006, 1561 a.a. - 1670 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00023345-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a spectrin repeat containing protein expressed in skeletal and smooth muscle, and peripheral blood lymphocytes, that localizes to the nuclear membrane. Mutations in this gene have been associated with autosomal recessive spinocerebellar ataxia 8, also referred to as autosomal recessive cerebellar ataxia type 1 or recessive ataxia of Beauce. Alternatively spliced transcript variants encoding different isoforms have been described. [provided by RefSeq]
Sequence: LRKIQQSVSEFEDKLAVPIKICSSATETYKVLQEHMDLCQALESLSSAITAFSASARKVVNRDSCVQEAAALQQQYEDILRRAKERQTALENLLAHWQRLEKELSSFLTWEspecificaciones
NP_892006 | |
Liquid | |
23345 | |
SYNE1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
8B/CPG2/DKFZp781J13156/FLJ30878/FLJ41140/KIAA0796/KIAA1262/KIAA1756/MYNE1/SCAR8 | |
SYNE1 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LRKIQQSVSEFEDKLAVPIKICSSATETYKVLQEHMDLCQALESLSSAITAFSASARKVVNRDSCVQEAAALQQQYEDILRRAKERQTALENLLAHWQRLEKELSSFLTW | |
RUO | |
SYNE1 | |
Wheat Germ (in vitro) | |
GST |