missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SLC11A1 Full-length ORF (AAH41787.1, 1 a.a. - 178 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00006556-P01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16170802

  • 508.00€ / 25 microgramos
Fecha estimada de envío: 17-07-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene is a member of the solute carrier family 11 (proton-coupled divalent metal ion transporters) family and encodes a multi-pass membrane protein. The protein functions as a divalent transition metal (iron and manganese) transporter involved in iron metabolism and host resistance to certain pathogens. Mutations in this gene have been associated with susceptibility to infectious diseases such as tuberculosis and leprosy, and inflammatory diseases such as rheumatoid arthritis and Crohn disease. Alternatively spliced variants that encode different protein isoforms have been described but the full-length nature of only one has been determined. [provided by RefSeq]

Sequence: MTGDKGPQRLSGSSYGSISSPTSPTSPGPQQAPPRETYLRNIESDLQAGAVAGFKLLWVLLWATVLGLLCQRLAARLGVVTGKDLGEVCHLYYPKVPRTVLWLTIELAIVGSDMQEVIGTAIAFNLLSAGRYHPSVPQLFRPGREQLLLLPPLTSPSQSLFYPAVPSEAGLLPCFPEM
Especificaciones

Especificaciones

AAH41787.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
45.6kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
LSH/NRAMP/NRAMP1
SLC11A1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
6556
SLC11A1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MTGDKGPQRLSGSSYGSISSPTSPTSPGPQQAPPRETYLRNIESDLQAGAVAGFKLLWVLLWATVLGLLCQRLAARLGVVTGKDLGEVCHLYYPKVPRTVLWLTIELAIVGSDMQEVIGTAIAFNLLSAGRYHPSVPQLFRPGREQLLLLPPLTSPSQSLFYPAVPSEAGLLPCFPEM
RUO
SLC11A1
Wheat Germ (in vitro)
GST
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales