missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SEMA4C Partial ORF (NP_060259, 536 a.a. - 635 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00054910-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: LLIQHVMTSDTSGICNLRGSKKVRPTPKNITVVAGTDLVLPCHLSSNLAHARWTFGGRDLPAEQPGSFLYDARLQALVVMAAQPRHAGAYHCFSEEQGAREspecificaciones
NP_060259 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LLIQHVMTSDTSGICNLRGSKKVRPTPKNITVVAGTDLVLPCHLSSNLAHARWTFGGRDLPAEQPGSFLYDARLQALVVMAAQPRHAGAYHCFSEEQGAR | |
RUO | |
SEMA4C | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
54910 | |
SEMA4C (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ20369/KIAA1739/M-SEMA-F/MGC126382/MGC126383/SEMACL1/SEMAF/SEMAI | |
SEMA4C | |
Recombinant | |
wheat germ expression system |